Order Entry
Export
ContactUsLinkComponent
(-)-Quinine hydrochloride dihydrate
Health
Irritant
(-)-Quinine hydrochloride dihydrate
Catalog # ROTH6225.1
Supplier:  Roth Carl
CAS Number:  
Health
Irritant
(-)-Quinine hydrochloride dihydrate
Catalog # ROTH6225.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :396.91 g/mol
Melting Pt:158…160 °C
Flash Pt:122 °C
EINECS:205-001-1
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:6119-47-7
Formula:C₂₀H₂₅ClN₂O₂·2H₂O
MDL Number:MFCD00151248