Order Entry
Export
Orders LinkContactUsLinkComponent
 

(+)-Cinchonine 99%

Irritant
(+)-Cinchonine 99%
Certificates
(+)-Cinchonine 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
  • CAS Number:118-10-5
  • Formula:C₁₉H₂₂N₂O
  • MDL Number:MFCD00064372
  • Molecular Weight:294.4 g/mol
  • Merck Index:13,02308
  • Melting Pt:255…265 °C
  • Density:1,2 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-234-6

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    255 to 265 °C
  • Loss on drying:
    ≤1.0 %
  • Sulfated ash:
    ≤0.1 %
  • Specific optical rotation:
    +251° to +269° (20 °C, 589 nm) (c=2, 0.2M HCl)
  • Titration with HClO4:
    98.5 to 101.5 %
  • Other alkaloids:
    ≤3 % (HPLC) Quinidine + hydroquinidine
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 49,70 €
    49,70 €
    1 * 25 g
       49,70 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 134,00 €
    134,00 €
    1 * 100 g
       134,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 453,00 €
    453,00 €
    1 * 500 g
       453,00 €