ContactUsLinkComponent
Rotary vane vacuum pump D 2.5 E
  181-2041
 :  LEYBOLD
undefined
Rotary vane vacuum pump D 2.5 E
  181-2041
 :  LEYBOLD
  • Description:
    Rotary vane pump TRIVAC D 2.5 E (220–240 V, 50 Hz; 230 V, 60 Hz)
  • Pk:
    1
Two-stage, oil-sealed vacuum pump based on the rotary vane principle. The oil injected into the suction chamber is used to seal, lubricate and cool. The intake branch and exhaust are fitted with small flanges. Typically used for mass spectrometers, electron microscopes, sterilisers, freeze-drying units, in chemistry and research laboratories, for the production of imaging tubes, in general vacuum technology and as a pre-vacuum pump for high-vacuum pumping systems.
  • Small and compact design, high degree of reliability, quiet operation
  • Environmentally friendly (low oil consumption, complies with EMC standards, IP 54-protected)
  • Higher performance thanks to a 3-stage gas ballast device, high water vapour tolerance
  • Safe and intelligent protection for the vacuum (hermetic seal), process quality (low oil backflow)
  • Suitable for continuous operation at 1000 mbar, low energy consumption
 : Delivery includes pump, dirt collecting sieve, oil filling (N 62 standard) — supplied separately, gas ballast device, cable with EU plug and handle.