Thermomixer, thermomixer compact
460-0019
- Orbit:3 mm
- Temperature control range:Ambient +4...99 °C
- Description:Thermomixer compact, UK-plug
- Model:Thermomixer compact
- Heating speed:~5 °C/min
- Heating power:90 W
- Incubating accuracy:±1 between 20 and 45 °C
±2 below 20 and above 45 °C - Mixing speed range:300 - 1400 min⁻¹
- Temperature setting:1...99 °C
- Weight:2,4 kg
- W×D×H:165×240×145 mm
- Pk:1
Thermomixer compact is ideal for mixing and temperature control of 24×1,5/2,0 ml microcentrifuge tubes. Applications include enzyme reactions at temperatures of 4 °C above ambient, transformation, denaturation of DNA, RNA and proteins, cultivation of bacteria and yeasts, bead technology, plasmid isolation and isolating DNA fragments from agarose gels.
- Dedicated compact unit for 24×1,5/2,0 ml microcentrifuge tubes
- Short mix function
- Quiet operation and high stability even at maximum mixing frequency
IP protection class according to DIN EN 61010: IP 20
: IP protection class according to DIN EN 61010: IP 20
: Supplied with a fixed thermoblock for 1,5/2,0 ml microcentrifuge tubes, IsoTherm rack and an IsoTherm cooling pack for sample storage at 0 °C.