Accessories for FlashGel™ system
Supplier: LONZA
About this item
13 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
- Catalog No:
- LONZ57068
- LONZ57069
- 733-1609
- 733-1616
- 733-1622
- 733-1621
- 733-1620
- 733-1734
- 733-1619
- LONZ57060
- 733-1623
- LONZ57064
- 733-1739
- Separation range (bp):
- 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1.2% agarose: 50-4000
2.2% agarose: 10-1000 - 1.2% agarose: 50-4000
2.2% agarose: 10-1000 - 1.2% agarose: 50-4000
2.2% agarose: 10-1000
- Storage (cassettes):
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Well volume:
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- Gel size W×D×H:
- 84x70x2 mm
- 84×70×2 mm
- 84x70x2 mm
- 84x70x2 mm
- 84×70×2 mm
- Cassette size W×D×H:
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- Dock size W×D×H:
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- LONZ57068FlashGel™ power supplySpecifications:
More
Specifications
Cat. No.LONZ57068Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - LONZ57069FlashGel™ power supply pack, includes FlashGel™ dockSpecifications:
More
Specifications
Cat. No.LONZ57069Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - 733-1609FlashGel™ dockSpecifications:
More
Specifications
Cat. No.733-1609Separation range (bp)Storage (cassettes)Well volume2 - 5 µlGel size W×D×H84x70x2 mmCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1616FlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettesSpecifications:
More
Specifications
Cat. No.733-1616Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - 733-1622FlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettesFlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettesLog in for availability1 * 500 µl320,00 €Specifications:
More
Specifications
Cat. No.733-1622Separation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlGel size W×D×H84×70×2 mmCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1621FlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applicationsFlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applicationsLog in for availability1 * 250 µl320,00 €Specifications:
More
Specifications
Cat. No.733-1621Separation range (bp)1.2% agarose: 50-4000
2.2% agarose: 10-1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlGel size W×D×H84x70x2 mmCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1620FlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettesFlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettesLog in for availability1 * 500 µl320,00 €Specifications:
More
Specifications
Cat. No.733-1620Separation range (bp)1.2% agarose: 50-4000
2.2% agarose: 10-1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlGel size W×D×H84x70x2 mmCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1734RNA marker (0,5 bp to 9 Kb)Specifications:
More
Specifications
Cat. No.733-1734Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - 733-1619FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)Log in for availability1 * 5 mL255,00 €Specifications:
More
Specifications
Cat. No.733-1619Separation range (bp)1.2% agarose: 50-4000
2.2% agarose: 10-1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlGel size W×D×H84×70×2 mmCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - LONZ57060FlashGel™ recovery buffer, 2× 500 μlSpecifications:
More
Specifications
Cat. No.LONZ57060Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - 733-1623RNA marker (0,5 - 9 kb), 50 µLSpecifications:
More
Specifications
Cat. No.733-1623Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - LONZ57064FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and maskFlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and maskLog in for availability1 * 1 KIT736,00 €Specifications:
More
Specifications
Cat. No.LONZ57064Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H - 733-1739Formaldehyde sample bufferSpecifications:
More
Specifications
Cat. No.733-1739Separation range (bp)Storage (cassettes)Well volumeGel size W×D×HCassette size W×D×HDock size W×D×H



