Orders LinkContactUsLinkComponent
Standard conical ground joints
Supplier:   Lenz Laborglas
Standard conical ground joints
Supplier:  Lenz Laborglas
Certificates
Standard conical ground joints
Standard conical ground joints
88 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Leakproof
  • Minimal requirement for joint lubrication
  • Joints cannot be misaligned
Socket and cone joints with fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving high resistance to temperature changes.
DIN 12242 Form E (Socket joints)
DIN 12242 Form B (Cone joints)
ISO 383
 : DIN 12242 Form E (Socket joints), DIN 12242 Form B (Cone joints), ISO 383.
Product Family Options
Sockets
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • - NS
    120 mm
    5/13 NS
    8 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 15,70 €
    15,70 €
    1 * 1 items
       15,70 €
  • - NS
    120 mm
    7/16 NS
    10 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 13,70 €
    13,70 €
    1 * 1 items
       13,70 €
  • - NS
    130 mm
    10/19 NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,80 €
    3,80 €
    1 * 1 items
       3,80 €
  • - NS
    130 mm
    12/21 NS
    15 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,30 €
    3,30 €
    1 * 1 items
       3,30 €
  • - NS
    130 mm
    14/23 NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,50 €
    3,50 €
    1 * 1 items
       3,50 €
  • - NS
    130 mm
    19/26 NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,60 €
    3,60 €
    1 * 1 items
       3,60 €
  • - NS
    130 mm
    24/29 NS
    28 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,70 €
    5,70 €
    1 * 1 items
       5,70 €
  • - NS
    130 mm
    29/32 NS
    33 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,70 €
    5,70 €
    1 * 1 items
       5,70 €
  • - NS
    130 mm
    34/35 NS
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 9,80 €
    9,80 €
    1 * 1 items
       9,80 €
  • - NS
    130 mm
    40/38 NS
    44 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 14,90 €
    14,90 €
    1 * 1 items
       14,90 €
  • - NS
    145 mm
    45/40 NS
    50 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 15,00 €
    15,00 €
    1 * 1 items
       15,00 €
  • - NS
    145 mm
    50/42 NS
    54 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 29,80 €
    29,80 €
    1 * 1 items
       29,80 €
  • - NS
    145 mm
    55/44 NS
    60 mm
    2,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 31,70 €
    31,70 €
    1 * 1 items
       31,70 €
  • - NS
    165 mm
    60/46 NS
    65 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 43,50 €
    43,50 €
    1 * 1 items
       43,50 €
  • - NS
    165 mm
    71/51 NS
    75 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 58,40 €
    58,40 €
    1 * 1 items
       58,40 €
  • - NS
    185 mm
    85/55 NS
    90 mm
    3,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 92,30 €
    92,30 €
    1 * 1 items
       92,30 €
Cones
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 5/13 NS
    115 mm
    - NS
    4 mm
    0,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 15,00 €
    15,00 €
    1 * 1 items
       15,00 €
  • 7/16 NS
    115 mm
    - NS
    6 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 8,00 €
    8,00 €
    1 * 1 items
       8,00 €
  • 10/19 NS
    115 mm
    - NS
    8 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,00 €
    4,00 €
    1 * 1 items
       4,00 €
  • 12/21 NS
    130 mm
    - NS
    11 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,10 €
    4,10 €
    1 * 1 items
       4,10 €
  • 14/23 NS
    130 mm
    - NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,90 €
    3,90 €
    1 * 1 items
       3,90 €
  • 19/26 NS
    130 mm
    - NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,50 €
    4,50 €
    1 * 1 items
       4,50 €
  • 24/29 NS
    130 mm
    - NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,60 €
    4,60 €
    1 * 1 items
       4,60 €
  • 29/32 NS
    130 mm
    - NS
    26 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,90 €
    4,90 €
    1 * 1 items
       4,90 €
  • 34/35 NS
    130 mm
    - NS
    30 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 9,00 €
    9,00 €
    1 * 1 items
       9,00 €
  • 40/38 NS
    145 mm
    - NS
    36 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 13,10 €
    13,10 €
    1 * 1 items
       13,10 €
  • 45/40 NS
    145 mm
    - NS
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 17,10 €
    17,10 €
    1 * 1 items
       17,10 €
  • 50/42 NS
    145 mm
    - NS
    46 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 31,40 €
    31,40 €
    1 * 1 items
       31,40 €
  • 55/44 NS
    145 mm
    - NS
    50 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 34,00 €
    34,00 €
    1 * 1 items
       34,00 €
  • 60/46 NS
    160 mm
    - NS
    54 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 40,50 €
    40,50 €
    1 * 1 items
       40,50 €
  • 71/51 NS
    180 mm
    - NS
    65 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 63,90 €
    63,90 €
    1 * 1 items
       63,90 €
  • 85/55 NS
    180 mm
    - NS
    80 mm
    3,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 111,00 €
    111,00 €
    1 * 1 items
       111,00 €
Cones with tip
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 7/16 NS
    115 mm
    - NS
    6 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 9,80 €
    9,80 €
    1 * 1 items
       9,80 €
  • 10/19 NS
    115 mm
    - NS
    8 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7,40 €
    7,40 €
    1 * 1 items
       7,40 €
  • 12/21 NS
    130 mm
    - NS
    11 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,20 €
    4,20 €
    1 * 1 items
       4,20 €
  • 14/23 NS
    130 mm
    - NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,30 €
    3,30 €
    1 * 1 items
       3,30 €
  • 19/26 NS
    130 mm
    - NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,40 €
    4,40 €
    1 * 1 items
       4,40 €
  • 24/29 NS
    130 mm
    - NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,60 €
    4,60 €
    1 * 1 items
       4,60 €
  • 29/32 NS
    130 mm
    - NS
    2,0 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,90 €
    4,90 €
    1 * 1 items
       4,90 €
  • 34/35 NS
    130 mm
    - NS
    30 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 10,50 €
    10,50 €
    1 * 1 items
       10,50 €
  • 40/38 NS
    145 mm
    - NS
    36 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 15,50 €
    15,50 €
    1 * 1 items
       15,50 €
  • 45/40 NS
    145 mm
    - NS
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 21,60 €
    21,60 €
    1 * 1 items
       21,60 €
  • 50/42 NS
    145 mm
    - NS
    46 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 33,20 €
    33,20 €
    1 * 1 items
       33,20 €
  • 55/44 NS
    160 mm
    - NS
    50 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,30 €
    35,30 €
    1 * 1 items
       35,30 €
  • 60/46 NS
    185 mm
    - NS
    54 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 37,30 €
    37,30 €
    1 * 1 items
       37,30 €
  • 71/51 NS
    185 mm
    - NS
    65 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 70,50 €
    70,50 €
    1 * 1 items
       70,50 €
  • 85/55 NS
    185 mm
    - NS
    80 mm
    3,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 115,00 €
    115,00 €
    1 * 1 items
       115,00 €
Long ground cones
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 5/20 NS
    115 mm
    - NS
    4 mm
    0,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 160,00 €
    160,00 €
    1 * 10 items
       160,00 €
  • 7/25 NS
    115 mm
    - NS
    6 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
  • 10/30 NS
    115 mm
    - NS
    8 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 68,90 €
    68,90 €
    1 * 10 items
       68,90 €
  • 12/32 NS
    140 mm
    - NS
    11 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 64,50 €
    64,50 €
    1 * 10 items
       64,50 €
  • 14/35 NS
    140 mm
    - NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,70 €
    5,70 €
    1 * 1 items
       5,70 €
  • 19/38 NS
    140 mm
    - NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 59,80 €
    59,80 €
    1 * 10 items
       59,80 €
  • 24/40 NS
    140 mm
    - NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 80,00 €
    80,00 €
    1 * 10 items
       80,00 €
  • 29/42 NS
    140 mm
    - NS
    26 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 8,30 €
    8,30 €
    1 * 1 items
       8,30 €
Sockets with special tube dimensions
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • - NS
    145 mm
    14/23 NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,00 €
    4,00 €
    1 * 1 items
       4,00 €
  • - NS
    160 mm
    14/23 NS
    16 mm
    1,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3,80 €
    3,80 €
    1 * 1 items
       3,80 €
  • - NS
    210 mm
    29/32 NS
    33 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,80 €
    5,80 €
    1 * 1 items
       5,80 €
  • - NS
    280 mm
    45/50 NS
    48 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 25,40 €
    25,40 €
    1 * 1 items
       25,40 €
  • - NS
    380 mm
    29/32 NS
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 18,60 €
    18,60 €
    1 * 1 items
       18,60 €
Cones with drip tip
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 19/26 NS
    120 mm
    - NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 90,20 €
    90,20 €
    1 * 10 items
       90,20 €
  • 24/29 NS
    130 mm
    - NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 90,50 €
    90,50 €
    1 * 10 items
       90,50 €
  • 29/32 NS
    130 mm
    - NS
    26 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 90,50 €
    90,50 €
    1 * 10 items
       90,50 €
Cones with short tip
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 14/23 NS
    120 mm
    - NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7,80 €
    7,80 €
    1 * 1 items
       7,80 €
  • 19/26 NS
    120 mm
    - NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 88,20 €
    88,20 €
    1 * 10 items
       88,20 €
  • 24/29 NS
    130 mm
    - NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 9,20 €
    9,20 €
    1 * 1 items
       9,20 €
  • 29/32 NS
    130 mm
    - NS
    26 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 98,30 €
    98,30 €
    1 * 10 items
       98,30 €
Long ground cones with tip
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 14/23 NS
    120 mm
    - NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 79,70 €
    79,70 €
    1 * 10 items
       79,70 €
  • 19/38 NS
    120 mm
    - NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 87,10 €
    87,10 €
    1 * 10 items
       87,10 €
  • 24/40 NS
    130 mm
    - NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 116,00 €
    116,00 €
    1 * 10 items
       116,00 €
  • 29/42 NS
    130 mm
    - NS
    26 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 123,00 €
    123,00 €
    1 * 10 items
       123,00 €
Cones with tip and special tube dimensions
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 29/32 NS
    300 mm
    - NS
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 29,10 €
    29,10 €
    1 * 1 items
       29,10 €
  • 34/35 NS
    300 mm
    - NS
    30 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 26,60 €
    26,60 €
    1 * 1 items
       26,60 €
  • 45/40 NS
    300 mm
    - NS
    40 mm
    2,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 28,10 €
    28,10 €
    1 * 1 items
       28,10 €
  • 60/46 NS
    375 mm
    - NS
    54 mm
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 48,40 €
    48,40 €
    1 * 1 items
       48,40 €
  • 71/51 NS
    500 mm
    - NS
    65 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 64,30 €
    64,30 €
    1 * 1 items
       64,30 €
Long ground sockets
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • - NS
    120 mm
    5/20 NS
    8 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 153,00 €
    153,00 €
    1 * 10 items
       153,00 €
  • - NS
    120 mm
    7/25 NS
    10 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 153,00 €
    153,00 €
    1 * 10 items
       153,00 €
  • - NS
    120 mm
    10/30 NS
    13 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 67,10 €
    67,10 €
    1 * 10 items
       67,10 €
  • - NS
    120 mm
    12/32 NS
    15 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 62,80 €
    62,80 €
    1 * 10 items
       62,80 €
  • - NS
    120 mm
    14/35 NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,70 €
    5,70 €
    1 * 1 items
       5,70 €
  • - NS
    120 mm
    19/38 NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,10 €
    6,10 €
    1 * 1 items
       6,10 €
  • - NS
    130 mm
    24/40 NS
    28 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 209,00 €
    209,00 €
    1 * 10 items
       209,00 €
  • - NS
    130 mm
    29/42 NS
    33 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 22,20 €
    22,20 €
    1 * 1 items
       22,20 €
Tubes with socket and cone
Product Information
  • Cone size
    Total length
    Socket size
    Tube ext.Ø
    Wall thickness
    Availability
    Price
  • 14/23 NS
    120 mm
    14/23 NS
    17 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 155,00 €
    155,00 €
    1 * 10 items
       155,00 €
  • 19/26 NS
    125 mm
    19/26 NS
    22 mm
    1,8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 160,00 €
    160,00 €
    1 * 10 items
       160,00 €
  • 29/32 NS
    130 mm
    14/23 NS
    26 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 163,00 €
    163,00 €
    1 * 10 items
       163,00 €
  • 29/32 NS
    150 mm
    29/32 NS
    33 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 18,70 €
    18,70 €
    1 * 1 items
       18,70 €