VWR® Labomeca®, Feet for rods
Supplier: VWR Collection
Jump to:
About this item
Mounting brackets for Labomeca® assemblies.
- Used for attaching rods to a fixed point of the laboratory: Floor, walls, ceiling, tables, shelves and furniture
- Supplied with wood screw, can be secured by using Labomeca® Allen key
Delivery information: Supplied with wood screw.
2 Options Available Below
Product Details & Documents
Mounting brackets for Labomeca® assemblies.
- Used for attaching rods to a fixed point of the laboratory: Floor, walls, ceiling, tables, shelves and furniture
- Supplied with wood screw, can be secured by using Labomeca® Allen key
Delivery information: Supplied with wood screw.