VWR® Pak Rak™ Plus, Loaders
Catalog # 613-0347
Supplier: VWR Collection
Specifications
- Colour code:Red
- Description:Pak Rak™ Plus Loader
- Environmentally Preferable:Low Manufacturing Impact,Sustainable Packaging
- For use with:0,1 - 1250 µl tips
- Pk:1
Specifications
About this item
Pak Rak instantly load empty racks with high-quality VWR® pipette tips.
- Reusable loaders produce up to 5 times less waste compared to manual refilling systems
- Hands or gloved hands never touch the tips
A simple downward push dispenses 96 tips into an 8x12 rack.
Ordering information: Refillable boxes/racks are required for the loader and are sold separately.