Order Entry
Export
Orders LinkContactUsLinkComponent
Pipette tray

Pipette tray

Pipette tray
Catalog # 612-1810
Supplier:   KARTELL
Pipette tray
Catalog # 612-1810
Supplier:  KARTELL
Pipette tray
Pipette tray

Pipette tray

Price
28,30 €
Log in for availability
1 * 1 items
Product Details & Documents

PVC.

  • Very useful workstation holder for items other than pipettes
  • For 7 pipettes diameter <20 mm or for 16 pipettes diameter <10 mm
  • For pipettes of all lengths

W×D×H: 283×216×40 mm

Documents

Specifications
  • Description:
    Pipette tray