Nunc-Immuno™ modules
About this item
PS. Nunc-Immuno™ modules have been developed for ELISA techniques where the demands of versatility require a different solution to the standard 96-well plate format. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
- With Nunc StarWell™ modules incubation time can be reduced by more than 50% or the sensitivity similarly increased
- Small numbers of samples can be processed more economically
- Available framed or unframed, modules remain in the frame when inverted
- Choice of well shape: Round (U), flat (F) or C-shaped (flat with curved edges)
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains. PolySorp™ surface has a high affinity for hydrophobic molecules. MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum. MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit www.vwr.com or contact your local VWR sales office for further information.
18 Options Available Below
Product Details & Documents
PS. Nunc-Immuno™ modules have been developed for ELISA techniques where the demands of versatility require a different solution to the standard 96-well plate format. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
- With Nunc StarWell™ modules incubation time can be reduced by more than 50% or the sensitivity similarly increased
- Small numbers of samples can be processed more economically
- Available framed or unframed, modules remain in the frame when inverted
- Choice of well shape: Round (U), flat (F) or C-shaped (flat with curved edges)
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains. PolySorp™ surface has a high affinity for hydrophobic molecules. MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum. MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit www.vwr.com or contact your local VWR sales office for further information.
Specifications for Product Family
Specifications
- Catalog No:
- 735-0054
- 735-0052
- 735-0050
- 735-0055
- 735-0053
- 735-0021
- 735-0015
- 735-0063
- 735-0042
- 735-0043
- 735-0048
- 737-0240
- 735-0126
- 735-0066
- 735-0020
- 735-0049
- 735-0046
- 735-0067
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- NAR Certificate of Retirement
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
Specifications
Product Family Options
Nunc-Immuno™ modules, individual strips
Product Information
- ColourRecommended working volumeAvailabilityPrice
- 735-0054MaxiSorp™, F8 wells per stripSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0054Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0052MaxiSorp™, F16 wells per stripSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0052Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0050MaxiSorp™, U16 wells per stripSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0050Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0055PolySorp™, F8 wells per stripSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0055Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0053PolySorp™, F16 wells per stripSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0053Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact
Nunc-Immuno™ modules, framed
Product Information
- ColourRecommended working volumeAvailabilityPrice
- 735-0021MaxiSorp™, C8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0021Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0015MaxiSorp™, C8×12 StarWell™ per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0015Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0063MaxiSorp™, C12×8 per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0063Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0042MediSorp™, F8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0042Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0043MultiSorp™, F8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0043Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0048MaxiSorp™, F8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0048Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 737-0240MaxiSorp™, F8×12 wells per frame (uncertified)MaxiSorp™, F8×12 wells per frame (uncertified)Clear350 µlLog in for availability1 * 120 items1 910,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.737-0240Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 735-0126MaxiSorp™, F16 (2×8 wells) ×6 per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0126Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0066MaxiSorp™, U8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0066Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0020PolySorp™, C8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0020Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0049PolySorp™, F8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0049Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0046PolySorp™, F16 (2×8 wells) ×6 wells per framePolySorp™, F16 (2×8 wells) ×6 wells per frameClear350 µlLog in for availability1 * 60 items425,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0046Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact - 735-0067PolySorp™, U8×12 wells per frameSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.735-0067Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No.CERT-IT000330)Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- 8-well strip caps, PE, sterile
Product Information
- Catalog #AvailabilityPrice
- 8-well strip caps, PE
Product Information
- Catalog #AvailabilityPrice
- Frames, PE