Industrial gloves, PX130 & PX140
Supplier: Marigold (Ansell)
Certificates
About this item
Seamless nylon gloves in a lightweight ergonomic design for highly dextrous tasks.
- Flat dipped polyurethane coating offers unrivalled grip in a variety of conditions
- Flexible fabric aids dexterity
- Ventilated back ensures that skin can breathe naturally
Certifications: EN 388 (4131); Cat. II
Specifications
- Catalog No:
- 112-2475
- 112-2474
- 112-2476
- 112-2481
- 112-2473
- 112-2472
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Palm PU coated
- Palm PU coated
- Palm PU coated
- Palm PU coated
- Palm PU coated
- Palm PU coated
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (4131)
- EN 388 (4131)
- EN 388 (4131)
- EN 388 (4131)
- EN 388 (4131)
- EN 388 (4131)