Orders LinkContactUsLinkComponent
Glass Adapters
Glass Adapters
Supplier:   ROBU GLASFILTER
Glass Adapters
Supplier:  ROBU GLASFILTER
Certificates
Glass Adapters
4 Options Available Below
Product Details & Documents
ROBU® Glass Adapter for Crucibles is designed to securely connect filter crucibles to other laboratory apparatus. Made from durable glass, it ensures a leak-free fit, offering reliable and efficient performance during filtration and chemical processes.
  • Leak-free connection
  • Durable glass construction
  • Resistant to heat and chemicals
  • Ensures secure attachment for crucibles
  • Compatible with various filtration setups
  • Easy to clean and reusable
  • Reliable performance in laboratory filtration processes
The ROBU Glass Adapter for Crucibles is designed to provide a secure and leak-free connection between filter crucibles and other laboratory apparatus. Constructed from durable, heat-resistant glass, this adapter is highly resistant to chemicals and ensures consistent, reliable performance during filtration processes. It is compatible with a wide range of filtration setups, offering versatility and ease of use in various laboratory applications. The adapter is easy to clean and reusable, making it a cost-effective solution for maintaining high standards in laboratory filtration work.
 : In compliance with ISO 4796-80 and ASTM E128-99 standards.
Product Family Options
Product Information
  • Material
    Ext.Ø
    Availability
    Price
  • 511-2108
    Glass adapter; inner tube Ø: 27 mm; outer tube Ø: 10 mm
    Glass adapter; inner tube Ø: 27 mm; outer tube Ø: 10 mm
    Borosilicate glass 3.3
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 24,60 €
    24,60 €
    1 * 1 items
       24,60 €
  • 511-2208
    Glass adapter; inner tube Ø: 34 mm; outer tube Ø: 10 mm
    Glass adapter; inner tube Ø: 34 mm; outer tube Ø: 10 mm
    Borosilicate glass 3.3
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 25,40 €
    25,40 €
    1 * 1 items
       25,40 €
  • 511-2308
    Glass adapter; inner tube Ø: 40 mm; outer tube Ø: 10 mm
    Glass adapter; inner tube Ø: 40 mm; outer tube Ø: 10 mm
    Borosilicate glass 3.3
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,00 €
    27,00 €
    1 * 1 items
       27,00 €
  • 511-2408
    Glass adapter; inner tube Ø: 50 mm; outer tube Ø: 10 mm
    Glass adapter; inner tube Ø: 50 mm; outer tube Ø: 10 mm
    Borosilicate glass 3.3
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 28,80 €
    28,80 €
    1 * 1 items
       28,80 €