Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boxes
Storage Boxes
Accessories for Transport and Storage Boxes
Accessories for Transport and Storage Boxes
Accessories for Transport and Storage Boxes
Catalog # 216-1775
Supplier:
LINPAC
Expand image
Accessories for Transport and Storage Boxes
Catalog # 216-1775
Supplier:
LINPAC
Specifications
Description:
Label attachment for all boxes
L×W:
- mm
Pk:
1
Specifications
About this item
Product Details
Product Details