Transport and Storage Boxes
Supplier: LINPAC
Jump to:
About this item
Plastic, white.
- With tightly fitting lid
- Suitable for use at −50 °C
Storage boxes made of polystyrene, white and lid made of soft polyethylene.
5 Options Available Below
Product Details & Documents
Plastic, white.
- With tightly fitting lid
- Suitable for use at −50 °C
Storage boxes made of polystyrene, white and lid made of soft polyethylene.
Product Family Options
Product Information
Accessories
Product Information
- Catalog #L×WAvailabilityPrice
- Label attachment for all boxes