Honey jars
Supplier: MUELLER KREMPEL
Certificates
About this item
Glass, clear, without lid.
- Good chemical and physical properties
- For use in applications without high thermal loads
- Metal screw cap available separately (see 216-3664 and 216-3664)
Perfect for sample collection, storage and transport of powders, pastes and liquids.