Safety basket
28 l, 470×340×250 mm
113-9051
HDPE.
- Two folding handles
- Space-saving, stackable
- Colors are not selectable
These safety baskets are designed for transport, storage and drainage of laboratory materials.
- Capacity:28 l
- Seq:2
- W×D×H:470×340×250 mm