ContactUsLinkComponent
Film, Kodak BioMax® MS
Film, Kodak BioMax® MS
Catalog # 730-3223
Supplier:  CARESTREAM HEALTH
undefined
Film, Kodak BioMax® MS
Catalog # 730-3223
Supplier:  CARESTREAM HEALTH
Specifications
  • Dimensions:
    350×430 mm
  • Seq:
    9
  • Pk:
    50
Specifications
About this item
BioMax® MS film is a high performance all-purpose, maximum sensitivity autoradiography film. When used with the BioMax® TranScreen HE intensifying screen it delivers four to eight times greater sensitivity in the detection of ³²P when compared to other autoradiography film and intensifying screen combinations. This film/screen combination is ideal for detection of Northern, Southern and Dot Blotting applications, particularly when detection of low intensity samples or exposure time reduction is required.
  • General purpose autoradiography film
  • Particularly recommended for ³²P-labelled samples
  • Suitable for manual or automated processing