Cooling spray
Cooling Spray
Catalog # 291-1203
Supplier: HM INDUSTRIESERVICE
Jump to:
Product Details & Documents
For applications in the vicinity of ignition sources or in potentially explosive areas. Used to detect thermal faults in electronics, to join small parts by means of cold shrinkage, to remove sticky dirt from textiles, to quick-freeze rubber and plastic and to test the function of thermostats and temperature probes.
- Cools down to −55 °C
- No risk of ignition or explosion, even in the presence of ignition sources
- Does not contain any CFCs
Documents
HMIN100089_TDS_EN
Datasheet
Specifications
- Description:Cooling Spray