Murexide
Glass bottle for solids, 25 g
Catalog # MOLEM78680857
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:13,06330
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



