Order Entry
Export
Orders LinkContactUsLinkComponent
1,2,4-Triazole

1,2,4-Triazole

Glass bottle for solids, 100 g
Catalog # MOLE53977204
Health
Irritant
Supplier:   Molekula
1,2,4-Triazole
Catalog # MOLE53977204
Supplier:  Molekula
CAS Number:  288-88-0
1,2,4-Triazole

Glass bottle for solids, 100 g

CAS Number:  288-88-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,2,4-1H-Triazole, 1H-1,2,4-Triazole
  • CAS Number:288-88-0
  • Formula:C₂H₃N₃
  • MDL Number:MFCD00005228
  • Molecular Weight:69.07 g/mol
  • Merck Index:12,09735
  • Boiling Pt:260 °C (1013 hPa)
  • Melting Pt:116…119 °C
  • Density:1.394 g/cm³ (25 °C)
  • EINECS:206-022-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications