1,2,3-Triazole
Glass bottle for solids, 1 g
Catalog # MOLEM18484185
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole
- CAS Number:288-36-8
- Formula:C₂H₃N₃
- MDL Number:MFCD00014490
- Molecular Weight:69.07 g/mol
- Boiling Pt:208 °C (1013 hPa)
- Melting Pt:23…25 °C
- Density:1,192 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



