

4-tert-Octylphenol
Catalog # EHERC15712100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:206.33 g/mol
Boiling Pt:279…281 °C (1013 hPa)
Melting Pt:80…81 °C
Density:0.932 g/cm³ (20 °C)
Flash Pt:147 °C
EINECS:205-426-2
UN:2430
ADR:8,III
:GHS05
:GHS09
CAS Number:140-66-9
Formula:C₁₄H₂₂O
MDL Number:MFCD00002368