Pararosaniline hydrochloride 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:569-61-9
:
Formula:C₁₉H₁₈ClN₃
MDL Number:MFCD00001657
:323.82 g/mol
Melting Pt:268…270 °C (dec.)
EINECS:209-321-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Green to dark green Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (110°C, 1 h)
- HPLC:≥97.5 %
- UV Lambda max:543 to 549 nm (c=0.0003%, 50% EtOH)