Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl cellulose
Methyl cellulose, 15 cP (2% H₂O), Sigma-Aldrich®
Methyl cellulose, 15 cP (2% H₂O), Sigma-Aldrich®
Methyl cellulose, 15 cP (2% H₂O), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Methyl cellulose, 15 cP (2% H₂O), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
9004-67-5
:
MDL Number:
MFCD00081763
:
M-Clarity™ quality level = MQ200
Storage Temperature:
Ambient