α-Hydroxy-o-tolyl-β-D-glucopyranoside
Catalog # MOLE11409633-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:286.28 g/mol
Merck Index:12,08476
Melting Pt:199…201 °C
Density:1.42 g/cm³ (20 °C)
EINECS:205-331-6
CAS Number:138-52-3
Formula:C₁₃H₁₈O₇
MDL Number:MFCD00006590