α-Hydroxy-o-tolyl-β-D-glucopyranoside ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:138-52-3
:
Formula:C₁₃H₁₈O₇
MDL Number:MFCD00006590
:286.28 g/mol
Merck Index:14,8324
Melting Pt:200…203 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-331-6