Cell culture dishes, Nunc™ UpCell™ surface
Supplier: Thermo Fisher Scientific
Certificates
About this item
With lid, sterile. Single-use vented dishes feature a temperature-responsive cell culture surface that is ideal for culture passaging, single-cell analyses, and cell transplantation research.
- Surface allows harvesting of cells with intact surface proteins
- High cell viability is achieved with minimal hands-on time and without physical force
- Cell surface proteins are preserved as no trypsinisation required
- Releases adherent cells by reduction of temperature of the cell culture
- Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix