Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Flunixin-5-hydroxy
Flunixin-5-hydroxy
EHERC13727100
:
EHRENSTORFER
New Product
:
Flunixin-5-hydroxy
EHERC13727100
:
EHRENSTORFER
New Product
:
Pk:
10 mg
CAS No.:
75369-61-8
Description:
Flunixin-5-hydroxy
Formula:
C₁₄H₁₁F₃N₂O₃
Storage temperature:
20 °C
Molecular weight (Mw):
312.24 g/mol