Cyprodinil-4'-hydroxy
100 µg/ml in Acetonitrile, 241.29 g/mol, 1 mL
Catalog # EHERA11909100AL-10
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
Certifications: This certificate is designed in accordance with ISO 17034 and ISO Guide 31. This certified reference material(CRM) was designed, produced and verified in accordance with ISO/IEC 17025, ISO 17034 and a registered quality management system ISO 9001.
Specifications
Specifications
- Concentration:100 µg/ml in Acetonitrile
- Pk:1 mL
- Molecular weight (Mw):241.29 g/mol
- Cat. No.:EHERA11909100AL-10
- Formula:C₁₄H₁₅N₃O
- Storage temperature:18 °C
- GHS signal:Danger
- CAS No.:195157-66-5



