Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Celite® 503, filter aid
Celite® 503, filter aid
Celite® 503, filter aid
Health
Celite® 503, filter aid
Catalog # ROTH0007.1
Supplier:
Roth Carl
CAS Number:
Expand image
Health
Celite® 503, filter aid
Catalog # ROTH0007.1
Supplier:
Roth Carl
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
2,5 kg
Seq:
4076
Specifications
About this item
Product Details
:
:
60.08 g/mol
:
GHS08
CAS Number:
68855-54-9
Formula:
O₂Si
Product Details