3,4-Diethoxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
Catalog # SIAL310778-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:170.16 g/mol
Boiling Pt:88…92 °C (0.4 mmHg)
Density:1.15 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
CAS Number:5231-87-8
Formula:C₈H₁₀O₄
MDL Number:MFCD00001333