3,5-Dinitro-o-toluamide, Sigma-Aldrich®
Catalog # SIAL524417-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:225.16
Merck Index:12,03321
Melting Pt:180 °C
:M-Clarity™ quality level = MQ100
EINECS:205-706-4
CAS Number:148-01-6
Formula:C₈H₇N₃O₅
MDL Number:MFCD00072065