Sudan I
Catalog # EHERC16986101
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:248.28 g/mol
Boiling Pt:444 °C (1013 hPa)
Melting Pt:135 °C
Density:1.119 g/cm³ (151 °C)
EINECS:212-668-2
CAS Number:842-07-9
Formula:C₁₆H₁₂N₂O
MDL Number:MFCD00003911