Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sudan III

Glass bottle for solids, 100 g
Catalog # ROTH4711.1
Supplier:   Roth Carl
Sudan III
Catalog # ROTH4711.1
Supplier:  Roth Carl
CAS Number:  85-86-9
Sudan III

Glass bottle for solids, 100 g

CAS Number:  85-86-9
Price
62,20 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Solvent Red 23, Sudan® III (Red), Scarlet B, Sudan Red III, Cerasin Red, Sudan Red BK, Sudan Red B, Fat Ponceau G, Sudan G, D and C Red No. 17, Oil red, Tony Red, Oil scarlet, Organol® red BS, 1-(4-(Phenylazo)phenylazo)-2-naphthol, Sudan V, 1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-ol
  • CAS Number:85-86-9
  • Formula:C₂₂H₁₆N₄O
  • MDL Number:MFCD00003905
  • Molecular Weight:352.4 g/mol
  • Merck Index:12,09054
  • Melting Pt:205.3…206.3 °C
  • EINECS:201-638-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications