Order Entry
Export
Orders LinkContactUsLinkComponent
Uric acid

Uric acid

Glass bottle for solids, 25 g
Catalog # ROTH4999.1
Supplier:   Roth Carl
Uric acid
Catalog # ROTH4999.1
Supplier:  Roth Carl
CAS Number:  69-93-2
Uric acid

Glass bottle for solids, 25 g

CAS Number:  69-93-2
Price
44,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1H-Purine-2,6,8(3H,7H,9H)-trione
  • CAS Number:69-93-2
  • Formula:C₅H₄N₄O₃
  • MDL Number:MFCD00005712
  • Molecular Weight:168.11 g/mol
  • Merck Index:12,10014
  • Melting Pt:436 °C
  • Density:1.89 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-720-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications