Uric acid
Glass bottle for solids, 25 g
Catalog # MOLE18110589-25G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione
- CAS Number:69-93-2
- Formula:C₅H₄N₄O₃
- MDL Number:MFCD00005712
- Molecular Weight:168.11 g/mol
- Merck Index:12,10014
- Melting Pt:436 °C
- Density:1.89 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-720-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



