Uric acid ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 69-93-2 Molecular Formula: C5H4N4O3 Formula Weight: 168.11 Melting Point: 300° Beilstein Reference: 156158 Merck Reference: 14,9875 MDL No.: MFCD00005712
Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione
- CAS Number:69-93-2
- Formula:C₅H₄N₄O₃
- MDL Number:MFCD00005712
- Molecular Weight:168.11 g/mol
- Merck Index:14,9875
- Melting Pt:>300 °C
- Density:1.89
- Storage Temperature:Ambient
- EINECS:200-720-7
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 69-93-2 Molecular Formula: C5H4N4O3 Formula Weight: 168.11 Melting Point: 300° Beilstein Reference: 156158 Merck Reference: 14,9875 MDL No.: MFCD00005712
Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione
- CAS Number:69-93-2
- Formula:C₅H₄N₄O₃
- MDL Number:MFCD00005712
- Molecular Weight:168.11 g/mol
- Merck Index:14,9875
- Melting Pt:>300 °C
- Density:1.89
- Storage Temperature:Ambient
- EINECS:200-720-7



