Order Entry
Export
ContactUsLinkComponent
2,3-Diaminonaphthalene
2,3-Diaminonaphthalene
Catalog # 00301.
Supplier:  Biotium
CAS Number:  
2,3-Diaminonaphthalene
Catalog # 00301.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :158.2 g/mol
Boiling Pt:343 °C (1013 hPa)
Boiling Pt:343 °C (1013 hPa)
Melting Pt:199 °C
Density:1,0968 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-241-0
CAS Number:771-97-1
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004116