1-Iodoperfluorohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:355-43-1
:
Formula:C₆F₁₃I
MDL Number:MFCD00001063
:445.95 g/mol
Boiling Pt:117 °C (1013 hPa)
Melting Pt:–45 °C
Density:2.0564 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:206-586-6