4-Chlorobenzhydrol
Glass bottle for solids, 100 mg
Catalog # PROOMM0380.09
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 4-Chlorophenyl phenyl carbinol, 4-Chlorobenzhydryl alcohol, 4-Chloro-α-phenylbenzyl alcohol, p-Chlorobenzhydrol, 4-Chlorodiphenylmethanol
- CAS Number:119-56-2
- Formula:C₁₃H₁₁ClO
- MDL Number:MFCD00004491
- Molecular Weight:218.68 g/mol
- EINECS:204-333-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



