Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4,5-diaminobenzotrifluoride
3-Bromo-4,5-diaminobenzotrifluoride
3-Bromo-4,5-diaminobenzotrifluoride
3-Bromo-4,5-diaminobenzotrifluoride
Supplier:
Thermo Scientific
Certificates
Expand image
3-Bromo-4,5-diaminobenzotrifluoride
Supplier:
Thermo Scientific
About this item
CAS Number:
113170-72-2
:
Formula:
C₇H₆BrF₃N₂
MDL Number:
MFCD00042484
:
255.04 g/mol
UN:
2811
ADR:
6.1,III