Nuclear Fast Red
Glass bottle, 1 L
Catalog # CTEH64242
Supplier: CLIN TECH
Jump to:
Product Details & Documents
Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
- CAS Number:6409-77-4
- Formula:C₁₄H₈NNaO₇S
- MDL Number:MFCD00078493
- Molecular Weight:357.28 g/mol
- Merck Index:13,06512
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:229-088-0
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



