Order Entry
Export
ContactUsLinkComponent
Nuclear Fast Red, Sigma-Aldrich®
Nuclear Fast Red, Sigma-Aldrich®
Catalog # 229113-5G
CAS Number:  
undefined
Nuclear Fast Red, Sigma-Aldrich®
Catalog # 229113-5G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance (Colour):
    Dark Brown to Very Dark Brown and Dark Red to Very Dark Red and Very Dark Red-Brown
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    515 - 540 nm
  • Extinction Coefficient
    c = 0.01 g/L, H₂O:
    ≥11000
  • Suitability
    Suitable for Determination of Calcium:
    Suitable

Specification Test Results

About this item

 :
 :357.28 g/mol
Merck Index:13,06512
Melting Pt:300 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:229-088-0
CAS Number:6409-77-4
Formula:C₁₄H₈NNaO₇S
MDL Number:MFCD00078493