Order Entry
Export
ContactUsLinkComponent
Sudan II, pure
Health
Sudan II, pure
Catalog # ACRO190150250
CAS Number:  
undefined
Health
Sudan II, pure
Catalog # ACRO190150250
CAS Number:  

Specification Test Results

  • Appearance:
    Red to orange-brown Powder
  • Infrared spectrum:
    Conforms
  • UV Lambda max:
    at approximately 420 and 493 nm (in methanol)
  • Specific extinction E (1%/1cm):
    ≥500 (at 493 nm, in methanol)

Specification Test Results

About this item