Order Entry
Export
Orders LinkContactUsLinkComponent
Spleen Dissociation Medium

Spleen Dissociation Medium

Spleen dissociation medium, 10 Vial
Catalog # STMC07915
Spleen Dissociation Medium
Catalog # STMC07915
CAS Number:  
Spleen Dissociation Medium
Spleen Dissociation Medium

Spleen dissociation medium, 10 Vial

CAS Number:  
Price
532,00 €
Log in for availability
1 * 10 Vial
Product Details & Documents

Medium for dissociation of mouse spleen.

  • Maximise recovery and viability of dendritic cells from mouse spleen
  • Optimised for use with selected EasySep™ cell separation kits

Maximise the recovery of dendritic cells from mouse spleens with Spleen dissociation medium, when used in combination with EasySep™ cell separation technology. This enzymatic dissociation medium contains collagenase, DNase, and fetal bovine serum (FBS), and has been optimised to maximise the viability of isolated spleen dendritic cells. Each 4 ml tube is sufficient for processing up to two mouse spleens.

 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    10 Vial
  • Description:
    Spleen dissociation medium
  • Cat. No.:
    STMC07915
  • Species:
    Mouse

Specifications