Fibronectin
Catalog # STMC07159
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 mg
- Species:Mouse
- Description:Fibronectin
Specifications
About this item
For coating tissue cultureware to promote cell adherence.
- Promote cell adherence on cultureware and coverslips
- Improve neural cell yield during in vitro differentiation of mouse embryonic stem cells
Support cell adherence on coverslips by using Fibronectin as a coating agent. In combination with DMEM/F-12 with 15 mM HEPES and ITS Supplement-A or ITS Supplement-B, this crucial glycoprotein can also be used to improve the yield of neural cells during the final stages of mouse embryonic stem (mES) cell in vitro differentiation into neural cell types.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').