Anti-FITC Monoclonal Antibody [clone: MOPC-21]
CAYM10343-100
- Clone:MOPC-21
- Isotype:IgG1
- Pk:100 Tests
Type: Control
Antigen: FITC
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Type: Control
Antigen: FITC
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: