Allyl cinnamate ≥98.0% stabilised
(stabilized with TBC)
CAS Number: 1866-31-5
MDL Number: MFCD00026105
Molecular Formula: C12H12O2
Molecular Weight: 188.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 156
Specific Gravity (20/20): 1.06
Product Categories: Materials Chemistry, Monomers, Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Cinnamic Acid Allyl Ester
: Allyl Cinnamate, Cinnamic Acid Allyl Ester, Cinnamic acid Allyl Ester
- :1866-31-5
- :C₁₂H₁₂O₂
- :MFCD00026105
- :188.23 g/mol
- :152 °C
- :1.06 (20 °C)
- :156 °C
- Purity/Analysis Method:>98.0% (GC)



