EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit
Catalog # STMC17861
Supplier: STEMCELL Technologies
Specifications
- Description:EasySep™ human Pan-CD25 positive selection and depletion Kit, 1×10⁹ cells
- Pk:1 KIT
Specifications
About this item
Immunomagnetic depletion of human CD25+ cells.
- Fast, easy-to-use and column-free
- Up to 1.3 log depletion of CD25+ cells
- Up to 98% purity
The EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit is designed to deplete CD25+ cells from fresh peripheral blood mononuclear cells or lysed leukapheresis samples. CD25+ cells are targeted with Tetrameric Antibody Complexes recognising CD25 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. labelled cells remain in the tube while unlabelled cells are poured off. The CD25 antigen is expressed on regulatory T cells and activated T and B cells.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').