(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl 95%
473232500.
New Product
:
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
- Appearance:white to light red powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Specific optical rotation:-56° to -59° (20°C, 589 nm) (c=1, Chloroform)
:
:285.35 g/mol
:Ambient
:3077
:9,III
:GHS05
:GHS09
:137848-29-4
:C₂₀H₁₅NO
:MFCD01882346